Orders LinkContactUsLinkComponent
Dipping bottles
Dipping bottles
Supplier:   Sampling Systems
Dipping bottles
Supplier:  Sampling Systems
Certificates
Dipping bottles
6 Options Available Below
Product Details & Documents
Dipping bottles are excellent for sampling in open water, as well as in tanks and drums.
  • Wide range available
  • Stainless steel 304 or brass construction
  • Customisable
Sampling liquids at depth is easy with these dipping bottles. Available in either 304 stainless steel or brass construction, they can take liquid samples from a variety of locations including open water kegs, drums, and wells.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • Brass
    300 ml
    162 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €
  • Brass
    500 ml
    240 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 308,00 €
    308,00 €
    1 * 1 ST
       308,00 €
  • Brass
    1000 ml
    300 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 336,00 €
    336,00 €
    1 * 1 ST
       336,00 €
  • Stainless steel 304
    300 ml
    153 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 461,00 €
    461,00 €
    1 * 1 ST
       461,00 €
  • Stainless steel 304
    500 ml
    207 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 510,00 €
    510,00 €
    1 * 1 ST
       510,00 €
  • Stainless steel 304
    1000 ml
    325 mm
    77 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 559,00 €
    559,00 €
    1 * 1 ST
       559,00 €