Inserts for U-D master manual samplers
Fournisseur: Sampling Systems
Certificats
À propos de cet article
Stainless steel inserts for U-D Master manual samplers.
- Made from 316 stainless steel
These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.
Frequently Bought Together