Orders LinkContactUsLinkComponent
V Spatula sampling tools
Supplier:   Sampling Systems
V Spatula sampling tools
Supplier:  Sampling Systems
Certificates
V Spatula sampling tools
V Spatula sampling tools
2 Options Available Below
Product Details & Documents
Ideal for taking surface samples of powders and viscous materials.
  • Multi-purpose sampling tool, available in a range of models
  • Suitable for sampling cheeses, waxes, and other hard materials
  • Robust V shaped blade allows for easy emptying
  • Made from chemically resistant stainless steel 316L
The V Spatula is available in varying models to offer the right solution. It is ideal for taking surface samples of a wide variety of substances, such as powders and harder materials such as waxes and cheeses, thanks to its robust blade design.
Product Family Options
Product Information
  • Material
    Length
    Availability
    Price
  • 300-0426
    V Spatula, 20 mm blade width
    V Spatula, 20 mm blade width
    Stainless steel 316L
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • 300-0427
    V Spatula, 20 mm blade width
    V Spatula, 20 mm blade width
    Stainless steel 316L
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €