Order Entry
Belgium
ContactUsLinkComponent
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Catalog # HONC34433-500ML
CAS Number:  
HYDRANAL™ - NEXTGEN Coulomat AG-FI, Anolyte for Coulometric Karl Fischer Titration, Fluka™
Catalog # HONC34433-500ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HYDRANAL™ - NEXTGEN Coulomat AG-FI
  • Seq:
    01

Specifications

About this item

Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).

  • Imidazole-free
  • Assay of water: 5 - 50 ppm
  • Methanol as solvent

The suffix “FI”, added to the name of the reagent, indicates that the reagent is “free of imidazole”. In 2015, the European Commission classified imidazole as a CMR substance and added H360D statement (“may damage the unborn child”).

Instead of imidazole, an alternative base has been used which does not compromise the performance of the Hydranal reagent.