ContactUsLinkComponent
Position TrueBlot® magnetic separator, 6
Position TrueBlot® magnetic separator, 6
Catalog # ROCKTMS-15-50
Position TrueBlot® magnetic separator, 6
Catalog # ROCKTMS-15-50
Specifications
  • Description:
    15/50 Tube TrueBlot® magnetic separator
  • Environmentally Preferable:
  • Shipping conditions:
    At ambient temperatures
  • Pk:
    1
Specifications
About this item
6x1,5 ml Position magnetic separator
  • Magnetic separation
  • Six 1,5 ml microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1,5 ml microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 ml.
Magnetic separator unit can be used to hold, and separate magnetic particles.