Order Entry
Belgium
ContactUsLinkComponent
Mouse IFNB1 / IFN-beta / Interferon beta Protein
Mouse IFNB1 / IFN-beta / Interferon beta Protein
Catalog # SINO50708-MCCH-100
Supplier:  Sino Biological
CAS Number:  
undefined
Mouse IFNB1 / IFN-beta / Interferon beta Protein
Catalog # SINO50708-MCCH-100
Supplier:  Sino Biological
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage conditions:
    –20…–80 °C
  • Endotoxin content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    Measured in antiviral assays using L929 cells infected with vesicular stomatitisvirus (VSV).
    The ED50 for this effect is 3 - 18 pg/ml.
  • Gene ID:
    P01575
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Ifb,IFN-beta,IFNB
  • Protein/peptide name:
    IFNB1
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant mouse IFNB1 consists of 166 amino acids and has a calculated molecular mass of 20.3 kDa. The recombinant protein migrates as an approximately 30 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Asn182
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

Specifications

About this item

A DNA sequence encoding the mouse IFNB1 (P01575) (Met 1-Asn 182) was fused with five amino acids (DDDDK) at the C-terminus was expressed and purified.