Orders LinkContactUsLinkComponent
Wire baskets, round, with handle
Wire baskets, round, with handle
Supplier:   RSG
Wire baskets, round, with handle
Supplier:  RSG
Certificates
Wire baskets, round, with handle
Wire baskets, round, with handle
2 Options Available Below
Product Details & Documents
Made from stainless steel 1.4301.
  • Rust-proof
  • Very good temperature resistance
  • Good chemical resistance
  • With reinforced edges
Product Family Options
Product Information
  • Ø×H
    Availability
    Price
  • 216-2376
    Wire basket, round, with handle
    Wire basket, round, with handle
    100×80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,50 €
    61,50 €
    1 * 1 ST
       61,50 €
  • 216-2377
    Wire basket, round, with handle
    Wire basket, round, with handle
    160×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 173,00 €
    173,00 €
    1 * 1 ST
       173,00 €