Order Entry
Belgium
ContactUsLinkComponent
Human recombinant DDR1 (from HEK293 cells), his tag
Human recombinant DDR1 (from HEK293 cells), his tag
Catalog # SINO10730-H08H-100
Supplier:  Sino Biological
CAS Number:  
undefined
Human recombinant DDR1 (from HEK293 cells), his tag
Catalog # SINO10730-H08H-100
Supplier:  Sino Biological
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20…−80 °C
  • Endotoxin content:
    <1,0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    NP_001945.3
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CAK,CD167,DDR,EDDR1,HGK2,MCK10,NEP,NTRK4,PTK3,PTK3A,RTK6,TRKE
  • Protein/peptide name:
    DDR1
  • Purity:
    >97% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant human DDR1 consists of 409 amino acids and has a calculated molecular mass of 45,7 kDa. The apparent molecular mass of rh DDR1 is approximately 55 - 60 kDa in SDS-PAGE under reducing conditions due to glycosylation.
  • Sequence:
    Met1-Thr416
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

Specifications

About this item

A DNA sequence encoding the extracellular domain of human DDR1 isoform b (NP_001945.3) (Met 1-Thr 416) was expressed, with a C-terminal polyhistidine tag.