Order Entry
Belgium
ContactUsLinkComponent
Human recombinant ERBB4 (from HEK293 cells), his tag
Human recombinant ERBB4 (from HEK293 cells), his tag
Catalog # SINO10363-H08H-50
Supplier:  Sino Biological
CAS Number:  
undefined
Human recombinant ERBB4 (from HEK293 cells), his tag
Catalog # SINO10363-H08H-50
Supplier:  Sino Biological
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Store it under sterile conditions at –20...−80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
  • Endotoxin content:
    <1,0 EU per μg of the protein as determined by the LAL method
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized human ErbB4-His at 10 μg/ml (100 μl/well) can bind biotinylated human NRG1 (Cat:11609 - H01H2), The EC50 of biotinylated human NRG1 (Cat:11609 - H01H2) is 0,4 - 0,92 μg/ml.
  • Gene ID:
    NP_005226.1
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Protein/peptide name:
    ERBB4
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant human ERBB4 comprises 635 amino acids and has a predicted molecular mass of 71,1 kDa. The apparent molecular mass of the protein is approximately 102 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Arg649
  • Formulation:
    Lyophilised from sterile PBS, pH 7,4.
  • Shipping temperature:
    In general, recombinant proteins are provided as lyophilized powder which are shipped at ambient temperature. Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.

Specifications

About this item

A DNA sequence encoding the human ERBB4 (NP_005226.1) (Met1-Arg649) with a C-terminal polyhistidine tag was expressed. Human and Rhesus ERBB4 sequences are identical.