Order Entry
Belgium
ContactUsLinkComponent
Human pro form of IL18 / Interleukin 18 / IGIF protein (GST Tag)(Inactive)
Human pro form of IL18 / Interleukin 18 / IGIF protein (GST Tag)(Inactive)
Supplier:  Sino Biological
Certificates
undefined
Human pro form of IL18 / Interleukin 18 / IGIF protein (GST Tag)(Inactive)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    GST Tag
  • Storage conditions:
    –20…−80 °C
  • Endotoxin content:
    Please contact us for more information.
  • Gene ID:
    Q14116
  • Reconstitution instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    IGIF,IL-18,IL-1g,IL1F4,Interleukin 18
  • Protein/peptide name:
    IL18
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular weight:
    The recombinant pro form of human IL18 consists of 419 amino acids and has a predicted molecular mass of 48,6 kDa.
  • Sequence:
    Met1-Asp193
  • Formulation:
    Powder
  • Shipping temperature:
    Ambient

Specifications

Frequently Bought Together