Human recombinant CCL1 (from HEK293 cells)
Catalog # SINO10057-H01H-20
Supplier: Sino Biological
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store it under sterile conditions at –20...–80 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin content:<1,0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_002972.1
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Protein/peptide name:CCL1
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:The recombinant human Fc/CCL1 is a disulfide-linked homodimeric protein. The reduced monomer consists of 310 amino acids and predicts a molecularmass of 35 kDa. As a result of glycosylation, the rhCCL1/Fc monomer migrates as an approximately 38 - 42 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Lys24-Lys96
- Formulation:Lyophilised from sterile PBS, pH 7,4
- Shipping temperature:In general, recombinant proteins are provided as lyophilised powder which are shipped at ambient temperature. Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.
Specifications
About this item
A DNA sequence encoding the mature form of human CCL1 (NP_002972.1) (Lys 24-Lys 96) was fused with the Fc region of human IgG1 at the N-terminus.