Order Entry
Belgium
ContactUsLinkComponent
 
Lysing Matrix E, MP Biomedicals
  116984001.
undefined
Lysing Matrix E, MP Biomedicals
  116984001.

 

  • Description:
    Lysing Matrix E
  • Adapter compatibility:
    96 Well Adapter
  • Instrument Compatibility:
    FastPrep-96
  • Tube compatibility:
    1,2 ml
  • Key applications:
    Extractions of nucleic acids, proteins, metabolites and other small molecules from soil, fecal, wastwater, sludge and other environmental samples
  • Application areas:
    Sample Preparation, Sample Lysis, Nucleic Acid Isolation & Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation
  • Material of construction:
    1,4 ceramic spheres, 0,1 mm silica spheres, and 4 mm glass beads.
  • Matrix type:
    E
  • Molecule enriched:
    DNA; RNA; Proteins; Other small molecules
  • Materials required (not supplied):
    FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • Storage and handling:
    Store at Room Temperature (15 to 30 °C)
  • Sample volume:
    up to 500 mg per prep
  • Sample type:
    Soil; Feces; Environmental Water; Wastewater; Sludge
  • Specific gravity:
    Ceramic Spheres, 4; Silica Spheres, 2,5; Glass Bead, 2,5
  • Product type:
    Lab Essentials
  • Pk:
    1

 

 

Lysing Matrix E is effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumours and other difficult tissues.

  • Impact resistant tube
  • Contains 1.4 mm ceramic spheres, 0.1 mm silica spheres and 4 mm glass beads
  • DNase and RNase Free

Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals receives the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.