Order Entry
Belgium
ContactUsLinkComponent
Lysing Matrix B, MP Biomedicals
Lysing Matrix B, MP Biomedicals
Catalog # 116540425.
Supplier:  MP Biomedicals
undefined
Lysing Matrix B, MP Biomedicals
Catalog # 116540425.
Supplier:  MP Biomedicals

Specifications

  • Description:
    Lysing Matrix B
  • Adapter compatibility:
    All FastPrep Adapters
  • Instrument Compatibility:
    FastPrep-24, FastPrep-96, SuperFastPrep-2, FastPrep-24 5G
  • Tube compatibility:
    2 ml
  • Key applications:
    Lysis of bacteria
  • Application areas:
    Nucleic Acid Isolation & Purification
  • Material of construction:
    0.1mm silica spheres
  • Molecule enriched:
    DNA, RNA, Proteins, Other small molecules
  • Materials required (not supplied):
    FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
  • Storage and handling:
    Store at Room Temperature (15 to 30° C)
  • Sample type:
    Bacteria (Gram +/-) and Spores
  • Specific gravity:
    2.5
  • Pk:
    250 g

Specifications

About this item

Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores

  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria

Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.