Anti-CASP11 Rat monoclonal antibody
NOVUNB120-10454
- Antibody type:Primary
- Antigen name:Caspase 11
- Antigen symbol:CASP11
- Clonality:Monoclonal
- Clone:17D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:12363
- Antigen synonyms:ich-3|Caspl|Casp11
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:A recombinant p30 subunit of mouse Caspase 11. [UniProt# P70343]
- Purification:Protein G purified
- Pk:0,1 mL
The Caspase-11 Antibody (17D9) from Novus Biologicals is a rat monoclonal antibody to Caspase-11. This antibody reacts with human, mouse. The Caspase-11 Antibody (17D9) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Microarray.
Microarray
Type: Primary
Antigen: Caspase 11
Clonality: Monoclonal
Clone: 17D9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse