Anti-L1CAM Mouse monoclonal antibody [clone: UJ127.11]
NOVUNB100-2682
- Antibody type:Primary
- Antigen name:L1 Cell Adhesion Molecule
- Antigen symbol:L1CAM
- Clonality:Monoclonal
- Clone:UJ127.11
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:3897
- Antigen synonyms:CD171|HSAS|CD171 antigen|HSAS1|S10|antigen identified by monoclonal R1|N-CAM-L1|neural cell adhesion molecule L1|SPG1|L1 cell adhesion molecule|CAML1N-CAML1|MASA|NCAM-L1|MIC5
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:Homogenous suspension of 16 week human foetal brain.
- Purification:IgG purified
- Pk:0,1 mg
Mouse Monoclonal L1CAM Antibody (UJ127.11). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: L1CAM
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human