Order Entry
Belgium
ContactUsLinkComponent
 
Anti-RNASET2 Rabbit Polyclonal Antibody
  NOVUNBP1-88753
Anti-RNASET2 Rabbit Polyclonal Antibody
  NOVUNBP1-88753

 

  • Antibody type:
    Primary
  • Antigen symbol:
    RNASET2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    8635
  • Antigen synonyms:
    EC 3.1.27.-|RNASE6PLbA514O12.3|FLJ42372|FLJ10907|ribonuclease T2|EC 3.1.27.1|Ribonuclease 6
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:FPNRSRFWKHEWEKHGTCAAQVDALNSQKKYFGRSLELYRELDLNSVLLKLGIKPSINYYQVADFKDALARVYGVIP
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μl
  • Pk:
    0,1 mL

 

 

The RNASET2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RNASET2. This antibody reacts with human. The RNASET2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RNASET2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human