ContactUsLinkComponent
LC-MS/MS system, SCIEX Triple Quad 4500
LC-MS/MS system, SCIEX Triple Quad 4500
Supplier:  AB SCIEX
Certificates
undefined
LC-MS/MS system, SCIEX Triple Quad 4500
Supplier:  AB SCIEX
Specifications
  • Catalog No:
  • 904-0091
  • 904-0019
  • Detector:
  • AcQuRate pµlse counting CEM
  • Mass range:
  • 5 - 2000 Da
  • Polarity switching:
  • 50 msec in MRM and schedµled MRM modes
  • Scan:
  • Q1 MS, Q3 MS, product ion, precursor ion, neutral loss or gain, MRM
Specifications
Front-End HPLC
Product Information
Frequently Bought Together