Order Entry
Belgium
Orders LinkContactUsLinkComponent
10X Tris-EDTA buffer pH 9,0

10X Tris-EDTA buffer pH 9,0

Tris-EDTA Buffer10X, 9,0, 500 mL
Catalog # NOVUNB900-62085
Supplier:   Novus Biologicals
10X Tris-EDTA buffer pH 9,0
Catalog # NOVUNB900-62085
Supplier:  Novus Biologicals
CAS Number:  
10X Tris-EDTA buffer pH 9,0

Tris-EDTA Buffer10X, 9,0, 500 mL

CAS Number:  
Price
599,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

10X Tris-EDTA buffer pH 9.0.

  • No preservative

Applications: Immunocytochemistry/Immunofluorescence
Immunohistochemistry
Immunohistochemistry-Paraffin

Storage temperature: Room temperature

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Tris-EDTA Buffer10X
  • pH value:
    9,0

Specifications