Orders LinkContactUsLinkComponent
Maybridge Screening library
Maybridge Screening library
Maybridge Screening library
Maybridge Screening library
The Maybridge portfolio consists of high-quality hit-like, lead-like and drug-like compounds which can generate valuable data in screening programs. They are developed specifically to support the pharmaceutical drug discovery process.
  • High-throughput screening for drug discovery
  • Maybridge hit-to-lead building blocks
  • Maybridge fragment libraries
See our dedicated Maybridge product page for more information and access to full library files, https://uk.vwr.com/cms/maybridge.
 : 96-well plates may be provided at mass ≥2 µmol or ≥0,5 mg as powder, dry films, or frozen DMSO solutions. 96- or 384-well plates may be provided at mass 0,1 µmol or 0,01 mg as dry films or frozen DMSO solutions. Compounds may also be ordered in vials at mass 2 µmol or 0,5 mg as powder or DMSO solutions.
We can also provide most major brands of plates and vials and we are happy to use those supplied by customers on request.
Product Family Options
Maybridge HitFinder screening library
  • Description
    Pk
  • Kit contents: 45 Plates, 0,25 µmol
    45
    1 * 45 ST
    1 * 45 ST
  • Kit contents: 180 plates, 1 µmol
    180
    1 * 180 ST
    1 * 180 ST
Maybridge HitCreater screening library
  • Description
    Pk
  • Kit contents: 45 Plates, 0,25 µmol
    45
    1 * 45 ST
    1 * 45 ST
  • Kit contents: 180 plates, 1 µmol
    180
    1 * 180 ST
    1 * 180 ST
Maybridge HitDiscover screening library
  • Description
    Pk
  • Kit contents: 652 plates, 1 µmol
    652
    1 * 652 ST
    1 * 652 ST
Screening library compounds, hand-selected
  • Description
    Pk
  • Kit contents: Hand-selected 96 well plate
    50
    1 * 50 ST
    1 * 50 ST
  • Kit contents: Hand-selected 96 well, 1 - 5 mg
    96
    1 * 96 ST
    1 * 96 ST
  • Kit contents: Hand-selected, 50×2 ml vial
    50
    1 * 50 ST
    1 * 50 ST