4-Hydroxy-4-methyl-2-pentanone ≥98.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:123-42-2
:
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471
:116.16 g/mol
EINECS:204-626-7
UN:1148
REACH:01-2119473975-21
:GHS02
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay:≥ 98,0 %
- Identification (IR) Passes test:Passes test
- Colouration ≤ 20 APHA:≤ 20 APHA
- Density (20/4):0,937 - 0,943
- Refractive index (20°C):1,422 - 1,425
- Evaporation residue:≤ 0,02 %
- Water:≤ 0,1 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC23427.290 spec_VWRC23427.320 spec_VWRC23427.460
Product specification
Frequently Bought Together