Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Catalog # 85783.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Catalog # 85783.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10 mg/kg
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Description:
    Ion chromatography anion multi-element standard solution (10 mg/kg each of: F⁻, Cl⁻, Br-, NO₃⁻, PO₄³⁻, SO₄²⁻ in H2O)
  • Storage temperature:
    Ambient

Specifications

About this item

This certified reference material is produced using a high purity starting material, acid from sub-boiling and 18 MOhm deionized water and filtered through a 0.2 micron filter.

10 mg/kg each of: F⁻, Cl⁻, Br-, NO₃⁻, PO₄³⁻, SO₄²⁻ in H2O

Certifications: CRM ISO 17034 and ISO 17025. Traceable to NIST.