ContactUsLinkComponent
Powered air respirator airflow indicator TR-970
Powered air respirator airflow indicator TR-970
Catalog # MMMATR-970
Supplier:  3M
undefined
Powered air respirator airflow indicator TR-970
Catalog # MMMATR-970
Supplier:  3M
Specifications
  • Description:
    Air flow indicator
  • Pk:
    1
Specifications
About this item
The TR-970 Air Flow Indicator is used to confirm that the manufacturer's minimum design flow rate of the Versaflo™ TR-300 Series Powered Air Turbo is exceeded.
  • Designed to enable testing at various elevations and temperatures within the operational range
  • Easy to use