ContactUsLinkComponent
Powered and supplied air replacement visors and lenses, Versaflo™
Powered and supplied air replacement visors and lenses, Versaflo™
Catalog # MMMAM927
Supplier:  3M
undefined
Powered and supplied air replacement visors and lenses, Versaflo™
Catalog # MMMAM927
Supplier:  3M
Specifications
  • Colour:
    Clear
  • Description:
    Replacement visors and lenses
  • Pk:
    1
Specifications
About this item
The Versaflo™ coated visor M-927 is a clear, polycarbonate replacement visor designed for use with 3M™ Versaflo™ M-Series Headtops.
  • Coated to provide chemical and scratch resistance
  • Clear polycarbonate
  • CE approved to EN 166 1:BT:3
Replace scratched and worn visors to ensure continued good vision. This replacement polycarbonate visor provides chemical and scratch resistance for 3M™ Versaflo™ M-Series Headtops.