ContactUsLinkComponent
Powered air respirator shrouds, Versaflo™
Powered air respirator shrouds, Versaflo™
Catalog # MMMAM448
Supplier:  3M
undefined
Powered air respirator shrouds, Versaflo™
Catalog # MMMAM448
Supplier:  3M
Specifications
  • Description:
    Powered air respirator shrouds
  • Pk:
    1
Specifications
About this item
The Versaflo™ Heavy Duty Shroud M-448 is a high durability shroud for 3M™ Versaflo™ M-400 helmets. A 3M™ Versaflo™ Inner Collar (M-444) must be zipped into the outer shroud before use.
  • Provides heavy-duty, durable protection
  • Designed for use with 3M™ Versaflo™ M-400 Helmets
Get added protection while wearing 3M™ Versaflo™ M-400 Helmets with the Versaflo™ Heavy Duty Shroud, M-448. A 3M™ Versaflo™ Inner Collar (M-444) must be zipped into the outer shroud before use.