ContactUsLinkComponent
Powered air respirator airflow deflector, Versaflo™
Powered air respirator airflow deflector, Versaflo™
N° de catalogue MMMAM-116
Fournisseur:  3M
undefined
Powered air respirator airflow deflector, Versaflo™
N° de catalogue MMMAM-116
Fournisseur:  3M
Spécifications
  • Colour:
    Blue
  • Description:
    Powered air respirator airflow deflector
  • Pk:
    1
Spécifications
À propos de cet article
The Versaflo™ Airflow Deflector M-116 is a replacement airflow deflector designed for use with 3M™ Versaflo™ M-100 and M-200 Series Headtops.
  • Replacement airflow deflector
Ensure airflow deflectors perform correctly with this blue, replacement 3M™ Versaflo™ Airflow Deflector, M-116. This airflow deflector is designed to be used as a spare part to replace worn or broken apparatus in 3M™ Versaflo™ M-100 and M-200 Series Headtops, and is CE approved.