Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide solution 10%, AnalaR NORMAPUR®
Corrosive
Sodium hydroxide solution 10%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sodium hydroxide solution 10%, AnalaR NORMAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay :
    10.0 - 10.5%
  • Heavy metals (as Pb):
    max. 5 ppm
  • Total nitrogen (N):
    max. 1 ppm
  • Carbonate (as Na₂CO₃):
    max. 1%
  • Chloride (Cl):
    max. 10 ppm
  • Phosphate (PO₄):
    max. 5 ppm
  • Silicate (as SiO₂):
    max. 10 ppm
  • Sulphate (SO₄):
    max. 5 ppm
  • Aluminium (Al):
    max. 5 ppm
  • Iron (Fe):
    max. 5 ppm

Specification Test Results

Related Information

Frequently Bought Together