Orders LinkContactUsLinkComponent
dNTP mix
dNTP mix
Supplier:   Biotium
dNTP mix
Supplier:  Biotium
Certificates
dNTP mix
2 Options Available Below
Product Details & Documents
Highly purified mix of dNTPs (>99% by HPLC) in 0,6 mM Tris-HCl (pH 7,5); each dNTP is at a concentration of 10 or 25 mm in the solution provided.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 40054.
    dNTP mix, 10 mm each, size: 5×1 ml
    dNTP mix, 10 mm each, size: 5×1 ml
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 482,00 €
    482,00 €
    1 * 5 ST
       482,00 €
  • 40053.
    dNTP mix, 25 mm each, size: 1 ml
    dNTP mix, 25 mm each, size: 1 ml
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.