Order Entry
Belgium
ContactUsLinkComponent
Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
Catalog # MDTC35-015-CF
Supplier:  Corning
undefined
Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
Catalog # MDTC35-015-CF
Supplier:  Corning

Specifications

  • Sterility:
    Sterile
  • Description:
    Fetal bovine serum, 50 ml
  • Origin:
    United States
  • Storage temperature:
    −40...−10°C
  • Pk:
    1

Specifications

About this item

Fetal bovine serum, United States origin collected at government approved abattoirs in the United States.

  • Shelf life: 60 months
  • Weight: 0,09 kg
  • Format: Liquid

True pooled and triple 0,1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.

Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).