Orders LinkContactUsLinkComponent
Masterflex Ultrapharm™ L/S® Custom-Engineered Panel-Mount Pumps and Controller, Avantor®
Masterflex Ultrapharm™ L/S® Custom-Engineered Panel-Mount Pumps and Controller, Avantor®
Masterflex Ultrapharm™ L/S® Custom-Engineered Panel-Mount Pumps and Controller, Avantor®
Certificates
Masterflex Ultrapharm™ L/S® Custom-Engineered Panel-Mount Pumps and Controller, Avantor®
Masterflex Ultrapharm™ L/S® Custom-Engineered Panel-Mount Pumps and Controller, Avantor®
4 Options Available Below
Product Details & Documents
Every drop counts—integrate into automated filling systems for high accuracy dosing and dispensing.
  • Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life
  • Masterflex double-Y tube sets ensure proper retention, repeatability, and accuracy
  • 250 rpm max speed; dosing volumes from <0.5 ml to >500 mL
  • Easy on tubing—planetary drive rollers reduce drag on tubing for longer tubing life and reduced risk of premature tubing failure
  • Whisper quiet operation reduces extra noise on the floor or in the lab
  • Autoclavable pump head; tested to 10 cycles without any loss of flow or accuracy performance
  • Pump head is available in either an aluminum body version or a 316 stainless steel body version; choose stainless steel for bioprocess and cleanroom applications
The Masterflex Ultrapharm™ L/S® Pump Head, part of the Masterflex® line of products, is designed for custom-engineered solutions to be used in the fill/finish step. This pump head produces gentle, low pulse, and highly accurate dosing and flow to ensure optimal integrity and uniformity. Designed with offset rollers, the pump head provides linear flow throughout the flow range. While one roller is drawing fluid through one leg of the double-Y tube set, the other roller is pushing fluid through the other leg. Also, dual channels allow for a lower rpm in order to achieve “single-channel” flow rates. A lower rpm means reduced shear forces to maintain fluid and molecular integrity.
Applications
Downstream process and fill/finish applications
Biopharma fill/finish for drug and vaccine production
High accuracy (±0.5%) dosing and dispensing
Pumping applications in cleanrooms or similar environments
Sanitary/sterile fluid dosing and dispensing
Specifications for Product Family
Specifications
  • Model:
  • 77313-20
  • 77313-22
  • 77313-30
  • 77313-32
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • Open-head sensor:
  • Yes
  • Yes
  • Yes
  • Yes
  • Optimum RPM range:
  • 250 rpm
  • 250 rpm
  • 250 rpm
  • 250 rpm
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • Tubing sizes accepted:
  • Double-Y L/S® 13, L/S® 14, L/S 16, L/S 25, L/S 17
  • Double-Y L/S® 15, L/S 24, L/S 35, L/S 36
  • Double-Y L/S® 13, L/S® 14, L/S 16, L/S 25, L/S 17
  • Double-Y L/S® 15, L/S 24, L/S 35, L/S 36
  • Volume per revolution with L/S 13 tubing:
  • 0,07 ml
  • 0,07 ml
  • Volume per revolution with L/S 14 tubing:
  • 0,25 ml
  • 0,25 ml
  • Volume per revolution with L/S 15 tubing:
  • 1,93 ml
  • 1,93 ml
  • Volume per revolution with L/S 16 tubing:
  • 0,86 ml
  • 0,86 ml
  • Volume per revolution with L/S 17 tubing:
  • 2,54 ml
  • 2,54 ml
  • Volume per revolution with L/S 24 tubing:
  • 2,87 ml
  • 2,87 ml
  • Volume per revolution with L/S 25 tubing:
  • 1,93 ml
  • 1,93 ml
  • Volume per revolution with L/S 35 tubing:
  • 3,99 ml
  • 3,99 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Specifications
Product Family Options
Product Information
  • Maximum Flow Rate
    Housing material
    Rotor material
    Availability
    Price
  • MFLX77313-20
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for Precision Tubing with Open-Head Sensor, Aluminum Housing, SS Rotor
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for Precision Tubing with Open-Head Sensor, Aluminum Housing, SS Rotor
    635 ml/min
    Aluminium
    Stainless Steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12 470,00 €
    12 470,00 €
    1 * 1 ST
       12 470,00 €
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77313-20
    Model
    77313-20
    No. of channels
    1
    Open-head sensor
    Yes
    Optimum RPM range
    250 rpm
    Pump series
    L/S
    Tubing sizes accepted
    Double-Y L/S® 13, L/S® 14, L/S 16, L/S 25, L/S 17
    Volume per revolution with L/S 13 tubing
    0,07 ml
    Volume per revolution with L/S 14 tubing
    0,25 ml
    Volume per revolution with L/S 15 tubing
    Volume per revolution with L/S 16 tubing
    0,86 ml
    Volume per revolution with L/S 17 tubing
    2,54 ml
    Volume per revolution with L/S 24 tubing
    Volume per revolution with L/S 25 tubing
    1,93 ml
    Volume per revolution with L/S 35 tubing
    Warranty
    2 years
    2 year warranty
  • MFLX77313-22
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for High-Performance Precision Tubing with Open-Head Sensor, Aluminum Housing, SS Rotor
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for High-Performance Precision Tubing with Open-Head Sensor, Aluminum Housing, SS Rotor
    998 ml/min
    Aluminium
    Stainless Steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12 470,00 €
    12 470,00 €
    1 * 1 ST
       12 470,00 €
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77313-22
    Model
    77313-22
    No. of channels
    1
    Open-head sensor
    Yes
    Optimum RPM range
    250 rpm
    Pump series
    L/S
    Tubing sizes accepted
    Double-Y L/S® 15, L/S 24, L/S 35, L/S 36
    Volume per revolution with L/S 13 tubing
    Volume per revolution with L/S 14 tubing
    Volume per revolution with L/S 15 tubing
    1,93 ml
    Volume per revolution with L/S 16 tubing
    Volume per revolution with L/S 17 tubing
    Volume per revolution with L/S 24 tubing
    2,87 ml
    Volume per revolution with L/S 25 tubing
    Volume per revolution with L/S 35 tubing
    3,99 ml
    Warranty
    2 years
    2 year warranty
  • MFLX77313-30
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for Precision Tubing with Open-Head Sensor, SS Housing, SS Rotor
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for Precision Tubing with Open-Head Sensor, SS Housing, SS Rotor
    635 ml/min
    Stainless steel
    Stainless Steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16 350,00 €
    16 350,00 €
    1 * 1 ST
       16 350,00 €
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77313-30
    Model
    77313-30
    No. of channels
    1
    Open-head sensor
    Yes
    Optimum RPM range
    250 rpm
    Pump series
    L/S
    Tubing sizes accepted
    Double-Y L/S® 13, L/S® 14, L/S 16, L/S 25, L/S 17
    Volume per revolution with L/S 13 tubing
    0,07 ml
    Volume per revolution with L/S 14 tubing
    0,25 ml
    Volume per revolution with L/S 15 tubing
    Volume per revolution with L/S 16 tubing
    0,86 ml
    Volume per revolution with L/S 17 tubing
    2,54 ml
    Volume per revolution with L/S 24 tubing
    Volume per revolution with L/S 25 tubing
    1,93 ml
    Volume per revolution with L/S 35 tubing
    Warranty
    2 years
    2 year warranty
  • MFLX77313-32
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for High-Performance Precision Tubing with Open-Head Sensor, SS Housing, SS Rotor
    Masterflex Ultrapharm™ L/S® OEM Panel-Mount Pump for High-Performance Precision Tubing with Open-Head Sensor, SS Housing, SS Rotor
    998 ml/min
    Stainless steel
    Stainless Steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16 350,00 €
    16 350,00 €
    1 * 1 ST
       16 350,00 €
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    MFLX77313-32
    Model
    77313-32
    No. of channels
    1
    Open-head sensor
    Yes
    Optimum RPM range
    250 rpm
    Pump series
    L/S
    Tubing sizes accepted
    Double-Y L/S® 15, L/S 24, L/S 35, L/S 36
    Volume per revolution with L/S 13 tubing
    Volume per revolution with L/S 14 tubing
    Volume per revolution with L/S 15 tubing
    1,93 ml
    Volume per revolution with L/S 16 tubing
    Volume per revolution with L/S 17 tubing
    Volume per revolution with L/S 24 tubing
    2,87 ml
    Volume per revolution with L/S 25 tubing
    Volume per revolution with L/S 35 tubing
    3,99 ml
    Warranty
    2 years
    2 year warranty
Accessories
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 14
    Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 14
    MFLX06451-14
    1,60 mm (0,063")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 102,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 25
    Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 25
    MFLX06451-25
    4,80 mm (0,189")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 134,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 16
    L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 16
    MFLX06456-16
    3,10 mm (0,122")
    PharmaPure®
    Log in for availability
    1 * 1 ST - 128,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 16
    Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 16
    MFLX06451-16
    3,10 mm (0,122")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 102,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 13
    Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 13
    MFLX06451-13
    0,80 mm (0,031")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 102,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 13
    L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 13
    MFLX06456-13
    0,80 mm (0,031")
    PharmaPure®
    Log in for availability
    1 * 1 ST - 116,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 14
    L/S® Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 14
    MFLX06456-14
    1,60 mm (0,063")
    PharmaPure®
    Log in for availability
    1 * 1 ST - 170,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 35
    High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 35
    MFLX06451-35
    7,90 mm (0,311")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 159,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 36
    High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 36
    MFLX06451-36
    9,70 mm (0,382")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 167,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 24
    L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 24
    MFLX06456-24
    6,40 mm (0,252")
    PharmaPure®
    Log in for availability
    1 * 1 ST - 191,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex® UltraPharm™ Pump Head, BioPharm Plus Platinum-Cured Silicone, L/S 24
    L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex® UltraPharm™ Pump Head, BioPharm Plus Platinum-Cured Silicone, L/S 24
    MFLX06445-24
    6,40 mm (0,252")
    BioPharm Plus platinum-cured silicone
    Log in for availability
    1 * 1 Voet - 224,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 15
    L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharmaPure®, L/S 15
    MFLX06456-15
    4,80 mm (0,189")
    PharmaPure®
    Log in for availability
    1 * 1 ST - 191,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 24
    High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 24
    MFLX06451-15
    4,80 mm (0,189")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 141,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 24
    High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, Puri-Flex®, L/S 24
    MFLX06451-24
    6,40 mm (0,252")
    Puri-Flex®
    Log in for availability
    1 * 1 ST - 1 010,00 €
Product Information
  • Catalog #
    Tubing int.Ø
    Tubing material
    Availability
    Price
  • L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharMed® BPT, L/S 36
    L/S® High-Performance Precision Pump Tubing, Double-Y Elements for Masterflex Ultrapharm™ Pump Head, PharMed® BPT, L/S 36
    MFLX06452-36
    9,70 mm (0,382")
    PharMed® BPT
    Log in for availability
    1 * 1 ST - 223,00 €