ContactUsLinkComponent
Masterflex® Single-Use Replacement Pump Heads with EZ-Set for Quattroflow™ Pump Systems, Avantor®
Masterflex® Single-Use Replacement Pump Heads with EZ-Set for Quattroflow™ Pump Systems, Avantor®
Certificates
undefined
Masterflex® Single-Use Replacement Pump Heads with EZ-Set for Quattroflow™ Pump Systems, Avantor®
Specifications
  • Model:
  • 76000-03
  • 76000-01
  • Inlet connection:
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp
  • Max. pressure:
  • 58 psi
  • 58 psi
  • Max. temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Outlet connection:
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp
  • Series:
  • Quattroflow
  • Quattroflow
  • Wetted materials:
  • Polypropylene (housing, valve plate), TPE (diaphragm), EPDM(valves, O-rings)
  • Polypropylene (housing, valve plate), TPE (diaphragm), EPDM(valves, O-rings)