ContactUsLinkComponent
CPC® High-Flow Plastic Quick-Disconnect Fittings, Hose Barb Inserts
CPC® High-Flow Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Certificates
undefined
CPC® High-Flow Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Specifications
  • Max. operating pressure:
  • 125 psi
  • 125 psi
  • Max. temperature:
  • 138 °C (280 °F)
  • 138 °C (280 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Seal material:
  • Platinum-Cured Silicone
  • Platinum-Cured Silicone
  • Sterilisation:
  • Autoclave or gamma
  • Autoclave or gamma
  • Warranty:
  • 1 year
  • 1 year