ContactUsLinkComponent
Reducer Fittings, Compression, Straight
Fitting, Brass, Straight, Compression Reducer, 3/8" OD x 1/4" OD
Catalog # MFLX31412-25
undefined
Fitting, Brass, Straight, Compression Reducer, 3/8" OD x 1/4" OD
Catalog # MFLX31412-25
Specifications
  • Connection Type:
    Compression Reducer
  • Flow pattern:
    Straight
  • Description:
    Fitting, Brass, Straight, Compression Reducer, ⅜" OD×¼" OD
  • Warranty:
    LTD life time
  • Pk:
    1
  • Fitting material:
    Brass
Specifications
About this item
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).
  • Connection Type:
    Compression Reducer
  • Flow pattern:
    Straight
  • Description:
    Fitting, Brass, Straight, Compression Reducer, ⅜" OD×¼" OD
  • Environmentally Preferable:
  • Warranty:
    LTD life time
  • Pk:
    1
  • Fitting material:
    Brass