ContactUsLinkComponent
Masterflex® Adapter Fittings, Luer to Luer, Elbow, Avantor®
undefined
Masterflex® Adapter Fittings, Luer to Luer, Elbow, Avantor®
  • Model:
  • 45508-84
  • 45508-26
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Connection type (port 2):
  • Male luer slip
  • Female luer
  • No. of ports:
  • 2
  • 2
Frequently Bought Together