ContactUsLinkComponent
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, PVDF, Avantor®
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, PVDF, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Tee, PVDF, Avantor®
Certificates
Specifications
  • Orifice dimensions:
  • 0,25"
  • 0,125"
  • 0,34375"
  • 0,125"
  • 0,5625"
  • 0,34375"
  • 0,125"
  • 0,25"
  • 0,125"
  • 0,34375"
  • 0,25"
  • 0,25"
  • 0,125"
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)