Orders LinkContactUsLinkComponent
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Certificates
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
4 Options Available Below
Product Details & Documents
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
Specifications for Product Family
Specifications
  • Inlet connection:
  • 1⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • Outlet connection:
  • 1⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • 1⁄4" Tri-Clamp
  • 3⁄4" Tri-Clamp
  • Wetted materials:
  • PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • 316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • 316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
  • Warranty:
  • Shelf life
  • Shelf life
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • MFLX76000-00
    Disposable Pump Chamber, PP machined for ABS and stainless steel single-use Quattroflow pump systems
    Disposable Pump Chamber, PP machined for ABS and stainless steel single-use Quattroflow pump systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 360,00 €
    1 360,00 €
    1 * 1 ST
       1 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX76000-00
    Inlet connection
    1⁄4" Tri-Clamp
    Outlet connection
    1⁄4" Tri-Clamp
    Wetted materials
    PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
    Warranty
    Shelf life
  • MFLX76000-02
    Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
    Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 310,00 €
    2 310,00 €
    1 * 1 ST
       2 310,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX76000-02
    Inlet connection
    3⁄4" Tri-Clamp
    Outlet connection
    3⁄4" Tri-Clamp
    Wetted materials
    PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
    Warranty
    Shelf life
  • MFLX76000-06
    Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
    Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 680,00 €
    2 680,00 €
    1 * 1 ST
       2 680,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX76000-06
    Inlet connection
    1⁄4" Tri-Clamp
    Outlet connection
    1⁄4" Tri-Clamp
    Wetted materials
    316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
    Warranty
    1 year
  • MFLX76000-08
    Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
    Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 110,00 €
    5 110,00 €
    1 * 1 ST
       5 110,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX76000-08
    Inlet connection
    3⁄4" Tri-Clamp
    Outlet connection
    3⁄4" Tri-Clamp
    Wetted materials
    316L SS (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
    Warranty
    1 year
Accessories
Product Information
  • Catalog #
    Electrical plug type
    Maximum Flow Rate
    Availability
    Price
  • Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VAC
    MFLX74000-49
    No plug
    3 L/min
    Log in for availability
    1 * 1 ST - 13 250,00 €