ContactUsLinkComponent
Masterflex® Adapter Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Masterflex® Adapter Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Certificates
Specifications
  • Max. temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Min. temperature:
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • –45 °C (–50 °F)
  • O-Ring material:
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sealable/weldable TPE
  • Sterilisation:
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Ethylene Oxide, Autoclave, and Gamme Irradiation
  • Warranty:
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect
  • Manufacturer defect