Orders LinkContactUsLinkComponent
Micropump® Eagle Drive™ Pump Systems
Micropump® Eagle Drive™ Pump Systems
Micropump® Eagle Drive™ Pump Systems
Micropump® Eagle Drive™ Pump Systems
Complete pumps include head, motor, and integrated drive.
  • Variable-speed electronic controller delivers smooth, pulseless flow
  • Static seals reduce maintenance
  • Small envelope-ideal for OEM use
Electromagnetically driven, precision-geared pumps provide smooth, accurate, pulseless flow. Torque improvements and compact size are achieved with NdFeB magnets. Control speed by using a 0 to 5 VDC input signal. Drives feature thermal and overload protection.
Choose a system with a suction shoe pump head to self compensate for wear, temperature, and pressure, and to maintain high volumetric efficiency. Cavity-style pump heads deliver higher lift capability and moderate pressure capability; an internal bypass is an integral safety feature of the cavity-style pump heads.
 : Includes 2-ft (0.6-m) power cord and screwdriver. Suction shoe service kits include: gears, suction shoe, shafts, scales, and springs. Cavity-style service kits include: gears, bushings, and seals.
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • Description
    Flow rate
    Maximum Flow Rate
  • Gear Pump Systems, Suction Shoe Pump Head, 316SS/PPS/PTFE, 0-5 VDC
    0,092 ml/rev
    460 ml/min
    1 * 1 ST
    1 * 1 ST - 3 410,00 €
    3 410,00 €
    1 * 1 ST
       3 410,00 €
    Specifications
    Cat. No.
    MFLX74215-10
    Warranty
    1 year
  • Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0-5 VDC
    0,32 ml/rev
    1,6 L/min
    1 * 1 ST
    1 * 1 ST - 3 050,00 €
    3 050,00 €
    1 * 1 ST
       3 050,00 €
    Specifications
    Cat. No.
    MFLX74445-05
    Warranty
    1 year
  • Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0,64 ml/rev
    0,64 ml/rev
    3,2 L/min
    1 * 1 ST
    1 * 1 ST - 3 120,00 €
    3 120,00 €
    1 * 1 ST
       3 120,00 €
    Specifications
    Cat. No.
    MFLX74445-15
    Warranty
    1 year
  • Gear Pump System, Cavity-style Pump Head, 316SS/PPS/PTFE, 0,91 ml/rev
    0,91 ml/rev
    4,55 L/min (1,2 gal./min)
    1 * 1 ST
    1 * 1 ST - 3 290,00 €
    3 290,00 €
    1 * 1 ST
       3 290,00 €
    Specifications
    Cat. No.
    MFLX74445-25
    Warranty
    1 year
Accessories
  •  
    Description
  • MFLX74445-91
    Gear Pump Service Kit, for 74445-05
    1 * 1 ST - 555,00 €