ContactUsLinkComponent
Masterflex® L/S® Tilt Bail for Digital Drives, Avantor®
Masterflex® L/S® Tilt Bail for Digital Drives, Steel-Plated
Catalog # MFLX07523-98
undefined
Masterflex® L/S® Tilt Bail for Digital Drives, Steel-Plated
Catalog # MFLX07523-98
Specifications
  • Description:
    Tilt Bail for L/S Digital Drives. 2 years warranty
  • Material:
    Plated steel
  • Pk:
    1
Specifications
About this item
Use to securely elevate front-end of digital drive.
  • Constructed with steel-plating for enhanced durability and corrosion resistance
Steel-plated elevating foot resists corrosion and comes with everything needed to mount onto your digital drive. Simple design elevates the front of the drive and swings out of the way when not in use.
Compatible with the following drives:
07522-20, 07552-30
07523-80, 07523-90
07528-10, 07528-20, 07528-30
07551-00, 07551-10
Compatible with the controller of the following modular drive:
77923-80
  • Description:
    Tilt Bail for L/S Digital Drives. 2 years warranty
  • Material:
    Plated steel
  • Environmentally Preferable:
  • Pk:
    1