ContactUsLinkComponent
Masterflex® Adapter Fittings, Luer to Luer, Straight, Avantor®
Masterflex® Adapter Fittings, Luer to Luer, Straight, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Luer to Luer, Straight, Avantor®
Certificates
Specifications
  • Model:
  • 30800-14
  • 45508-82
  • 45508-80
  • 45508-22
  • Connection type (port 1):
  • Male luer
  • Female luer
  • Female luer
  • Female luer
  • Connection type (port 2):
  • Male luer
  • Male luer lock
  • Male luer lock
  • Female luer
  • No. of ports:
  • 1
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months