Cadence Science Adapter Fittings, Luer Stopcock
Supplier: Avantor Fluid Handling
Certificates
About this item
Exceptional durability.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
- Full material traceability on all items
Easily connect your tubing to your system with these durable luer stopcocks.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX41607-02
- MFLX41607-00
- Connection type (port 1):
- Female luer
- Female luer
- Connection type (port 2):
- Female luer
- Male luer lock
- Connection type (port 3):
- Male luer lock
- No. of ports:
- 3
- 2