ContactUsLinkComponent
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Certificates
undefined
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Certificates
Specifications
  • Model:
  • 07624-30
  • 07624-28
  • 07624-26
  • 07624-48
  • 07624-46
  • 07624-38
  • 07624-36
  • 07624-34
  • 07624-32
  • 07624-18
  • 07624-24
  • 07624-44
  • 07624-22
  • 07624-42
  • 07624-40
  • Regulatory approvals:
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • 3A;ADCF;European Pharmacopoeia;FDA;Non-DEHP;RoHS;USP
  • Series:
  • Similar to SC0110
  • Similar to SC0109
  • Similar to SC0108
  • Similar to SC0119
  • Similar to SC0118
  • Similar to SC0114
  • Similar to SC0113
  • Similar to SC0112
  • Similar to SC0111
  • Similar to SCO107
  • Similar to SC0117
  • Similar to SC0106
  • Similar to SC0116
  • Similar to SC0115
  • Tubes per pack:
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • 6
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year