ContactUsLinkComponent
Masterflex® Check Valves, PTFE, Avantor®
Masterflex® Check Valves, PTFE, Avantor®
Certificates
undefined
Masterflex® Check Valves, PTFE, Avantor®
Specifications
  • Model:
  • 98515-14
  • 98515-12
  • 98515-18
  • Max. pressure:
  • 50 psi
  • 50 psi
  • 50 psi
  • Max. temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Orifice:
  • 6,35 mm (0,25")
  • 3,175 mm (0,125")
  • 12,7 mm (0,5")
  • Warranty:
  • 1 year
  • 1 year
  • 1 year