ContactUsLinkComponent
Masterflex® ACR Valves for Chemical and Corrosive Fluids, PTFE, Avantor®
Masterflex® ACR Valves for Chemical and Corrosive Fluids, PTFE, Avantor®
Certificates
undefined
Masterflex® ACR Valves for Chemical and Corrosive Fluids, PTFE, Avantor®
Specifications
  • Model:
  • 98516-18
  • 98516-02
  • 98516-10
  • 98516-12
  • Int. Ø tubing:
  • 0,25"
  • Max. temperature:
  • 156 °C (240 °F)
  • 156 °C (240 °F)
  • 156 °C (240 °F)
  • 156 °C (240 °F)
  • Orifice:
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 9,525 mm (0,375")
  • Tubing ext.Ø:
  • 0,25"
  • 0,375"
  • Wetted materials:
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year