Order Entry
Belgium
ContactUsLinkComponent
Masterflex® Touch-Screen Syringe Pumps, Avantor®
Masterflex® Touch-Screen Syringe Pumps, Avantor®
Certificates
undefined
Masterflex® Touch-Screen Syringe Pumps, Avantor®

Specifications

  • Accuracy:
  • ±0,355%
  • ±0,355%
  • ±0,355%
  • ±0,355%
  • ±0,355%
  • Linear force:
  • 75 lbs.
  • 75 lbs.
  • 75 lbs.
  • 75 lbs.
  • 75 lbs.
  • Output:
  • RS-232, RS-485, USB, ; 15 pin D-sub connector
  • RS-232, RS-485, USB, ; 15 pin D-sub connector
  • RS-232, RS-485, USB, ; 15 pin D-sub connector
  • RS-232, RS-485, USB, ; 15 pin D-sub connector
  • RS-232, RS-485, USB, ; 15 pin D-sub connector
  • Reproducibility:
  • ±0,05%
  • ±0,05%
  • ±0,05%
  • ±0,05%
  • ±0,05%
  • Syringe size accepted:
  • 0,5 µl - 140 ml
  • 0,5 µl - 140 ml
  • 0,5 µl - 140 ml
  • 0,5 µl - 140 ml
  • 0,5 µl - 140 ml
  • Syringe travel rate:
  • <190,8"
  • <190,8"
  • <190,8"
  • <190,8"
  • <190,8"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year

Specifications

Frequently Bought Together