ContactUsLinkComponent
Masterflex® Turbo-Prop Open Channel Flowmeters for Water, Avantor®
Masterflex® Turbo-Prop Open Channel Flowmeters for Water, Avantor®
Certificates
undefined
Masterflex® Turbo-Prop Open Channel Flowmeters for Water, Avantor®
Specifications
  • Model:
  • 32922-01
  • 32922-02
  • Accuracy:
  • ±0,1 ft/sec (average velocity)
  • ±0,1 ft/sec (average velocity)
  • Battery life:
  • 5 years
  • 5 years
  • Display type:
  • LCD with 4 button operation
  • LCD with 4 button operation
  • Flow Tube material:
  • PVC and anodized aluminum with SS water bearing
  • Max. operating temperature:
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • Warranty:
  • 1 year
  • 1 year