Order Entry
Belgium
ContactUsLinkComponent
Masterflex® B/T® Variable Speed and Air-Powered Analog Drives, Avantor®
Masterflex® B/T® Variable Speed and Air-Powered Analog Drives, Avantor®
Certificates
undefined
Masterflex® B/T® Variable Speed and Air-Powered Analog Drives, Avantor®

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Model:
  • 77711-95
  • 77111-80
  • 77111-67
  • No. of channels:
  • 1
  • 1
  • No. of rollers:
  • 3
  • 3
  • Control type:
  • Analog
  • Analog
  • Analog
  • Min. flow rate per channel:
  • 710 ml/min
  • Max. flow rate per channel:
  • 42000 ml/min
  • IP rating:
  • IP 56
  • IP 56
  • IP 56
  • Anti-drip:
  • No
  • Cloud connected:
  • No
  • Digital dispensing:
  • No
  • Min. RPM:
  • 35 rpm
  • 12 rpm
  • Motor:
  • 560 W
  • 370 W
  • Motor hp:
  • 3/4
  • 1/2
  • Open-head sensor:
  • No
  • Pump series:
  • B/T
  • B/T
  • B/T
  • Reversible motor:
  • No
  • Yes
  • Tubing sizes accepted:
  • B/T 87 or B/T 91
  • B/T 87 or B/T 91
  • Washdown capable:
  • Yes
  • Yes
  • Yes
  • Current:
  • 3,3 A
  • Frequency:
  • 50/60 Hz
  • Voltage:
  • 115 VAC
  • 230 VAC
  • Warranty:
  • 3 Months
  • 2 years
  • 2 years

Specifications

Related Information

Frequently Bought Together