ContactUsLinkComponent
Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
Certificates
undefined
Masterflex® B/T® Rapid-Load Pump Heads, Avantor®
Specifications
  • Model:
  • 77111-50
  • 77111-55
  • Gear ratio:
  • 5,6:1
  • 5,6:1
  • IP rating:
  • IP 56
  • IP 56
  • Motor mount:
  • NEMA Type 56C
  • IEC 72-71-14F130 with B5 160-mm flange
  • Mounting type:
  • Direct-coupled
  • Direct-coupled
  • No. of rollers:
  • 3
  • 3
  • Open-head sensor:
  • No
  • No
  • Pump series:
  • B/T
  • B/T
  • Tubing sizes accepted:
  • B/T 87 or B/T 91
  • B/T 87 or B/T 91
  • Volume per revolution with B/T 87 tubing:
  • 0,059 L (0,0156 gal.)
  • 0,059 L (0,0156 gal.)
  • Volume per revolution with B/T 91 tubing:
  • 0,131 L (0,035 gal.)
  • 0,131 L (0,035 gal.)
  • Warranty:
  • 2 years
  • 2 years