Orders LinkContactUsLinkComponent
Masterflex® Motorized Drum Pump Systems, Avantor®
Masterflex® Motorized Drum Pump Systems, Avantor®
Masterflex® Motorized Drum Pump Systems, Avantor®
Certificates
Masterflex® Motorized Drum Pump Systems, Avantor®
Masterflex® Motorized Drum Pump Systems, Avantor®
3 Options Available Below
Product Details & Documents
Complete drum pump system specifically packaged for different applications.
  • Flow rate 35 GMP for all models
  • Equipped with variable-speed control motors for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Pre-configured drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Based upon the application, polypropylene (PP), CPVC, PVDF, and 316 stainless steel (SS) materials provide excellent chemical resistance against corrosive liquids.
The Measurement Pump Packages (Models 70614-06, -08 and -09) include a digital flowmeter and totalizer. The dispensing hose for all the pumps is a 6-ft length of PVC tubing. The storage bracket for all packages is made from 16 gauge coated steel.
Delivery information: Includes electric motor with 16-ft (4.8-m) power cord, pump tube, wall storage bracket, barrel adapter, dispensing hose, and nozzle.
Specifications for Product Family
Specifications
  • Model:
  • 70614-00
  • 70614-02
  • 70614-03
  • 70614-04
  • 70614-05
  • 70614-06
  • 70614-09
  • 70614-10
  • Tube length, inlet:
  • 39"
  • 39"
  • 39"
  • 39"
  • 39"
  • 39"
  • 39"
  • 39"
  • Wetted materials:
  • CPVC
  • Polypropylene
  • Polypropylene
  • PVDF
  • PVDF
  • Polypropylene
  • PVDF
  • Stainless Steel
  • Warranty:
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years
  • 3 years
Specifications
Product Family Options
Product Information
  • Maximum Flow Rate
    Power
    Availability
    Price
  • MFLX70614-00
    Motorized Drum Pump Package, 35 gal./min, 39"L CPVC Tube; 115V
    Motorized Drum Pump Package, 35 gal./min, 39"L CPVC Tube; 115V
    35 gal./min
    115 VAC; 60 Hz
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX70614-00
    Model
    70614-00
    Tube length, inlet
    39"
    Wetted materials
    CPVC
    Warranty
    3 years
  • MFLX70614-02
    Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 115V
    Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 115V
    35 gal./min
    115 VAC; 50 Hz
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX70614-02
    Model
    70614-02
    Tube length, inlet
    39"
    Wetted materials
    Polypropylene
    Warranty
    3 years
  • MFLX70614-03
    Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 230V
    Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 230V
    35 gal./min
    230 VAC; 60 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 370,00 €
    2 370,00 €
    1 * 1 ST
       2 370,00 €
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70614-03
    Model
    70614-03
    Tube length, inlet
    39"
    Wetted materials
    Polypropylene
    Warranty
    3 years
    CE Compliant
  • MFLX70614-04
    Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 115V
    Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 115V
    35 gal./min
    115 VAC; 60 Hz
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX70614-04
    Model
    70614-04
    Tube length, inlet
    39"
    Wetted materials
    PVDF
    Warranty
    3 years
  • MFLX70614-05
    Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 230V
    Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 230V
    35 gal./min
    230 VAC; 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 580,00 €
    3 580,00 €
    1 * 1 ST
       3 580,00 €
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70614-05
    Model
    70614-05
    Tube length, inlet
    39"
    Wetted materials
    PVDF
    Warranty
    3 years
    CE Compliant
  • MFLX70614-06
    Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PP Tube; 115V
    Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PP Tube; 115V
    35 gal./min
    115 VAC; 60 Hz
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX70614-06
    Model
    70614-06
    Tube length, inlet
    39"
    Wetted materials
    Polypropylene
    Warranty
    3 years
  • MFLX70614-09
    Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PVDF Tube; 230V
    Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PVDF Tube; 230V
    35 gal./min
    230 VAC; 50 Hz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 500,00 €
    7 500,00 €
    1 * 1 ST
       7 500,00 €
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLX70614-09
    Model
    70614-09
    Tube length, inlet
    39"
    Wetted materials
    PVDF
    Warranty
    3 years
    CE Compliant
  • MFLX70614-10
    Motorized Drum Pump System, 35 gal./min, 39"L SS Tube; 115V
    Motorized Drum Pump System, 35 gal./min, 39"L SS Tube; 115V
    35 gal./min
    115 VAC; 60 Hz
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX70614-10
    Model
    70614-10
    Tube length, inlet
    39"
    Wetted materials
    Stainless Steel
    Warranty
    3 years