ContactUsLinkComponent
Masterflex® ADCF Polypropylene Sanitary Adapters, Avantor®
Masterflex® Sanitary End Cap, ADCF Polypropylene; 1-1/2", Avantor®
Catalog # MFLX31513-18
CE certificate
undefined
Masterflex® Sanitary End Cap, ADCF Polypropylene; 1-1/2", Avantor®
Catalog # MFLX31513-18
CE certificate
Specifications
  • Connection Type:
    Sanitary Cap
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    1-½"
  • Description:
    Sanitary End Cap, ADCF Polypropylene; 1-½"
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Pk:
    10
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    1-½"
Specifications
About this item
These adapters are perfect to transition from sanitary clamp connections to a barb for flexible tubing.
  • Constructed of durable Polypropylene
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI. Available with 3/8" or 1/2" hose barbs and 3/4" mini or 1-1/2" standard sanitary clamp connections.
  • Connection Type:
    Sanitary Cap
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    1-½"
  • Description:
    Sanitary End Cap, ADCF Polypropylene; 1-½"
  • Environmentally Preferable:
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Pk:
    10
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    1-½"