ContactUsLinkComponent
Kobold Industrial Flow Indicators with Plastic Rotor
undefined
Kobold Industrial Flow Indicators with Plastic Rotor
  • Max. pressure:
  • 230 psi
  • 230 psi
  • 230 psi
  • 230 psi
  • 230 psi
  • Max. operating temperature:
  • 98,9 °C (210 °F)
  • 98,9 °C (210 °F)
  • 98,9 °C (210 °F)
  • 98,9 °C (210 °F)
  • 98,9 °C (210 °F)
  • Pressure drop at max. flow:
  • 3,63 psid
  • 3,63 psid
  • 3,63 psid
  • 2,9 psid
  • 3,63 psid
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together